Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CK357656.1 | - | 1 - 2 | 216 | 75 | |
FQ217150.1 | - | 1 - 2 | 246 | 75 | ||
FQ217655.1 | - | 1 - 2 | 248 | 75 | ||
FQ220032.1 | - | 505 | 75 | |||
FQ221038.1 | - | 1 - 2 | 245 | 75 | ||
FQ221373.1 | - | 1 - 2 | 246 | 75 | ||
FQ222277.1 | - | 1 - 2 | 159 | 75 | ||
FQ222316.1 | - | 1 - 2 | 245 | 75 | ||
FQ222356.1 | - | 1 - 2 | 246 | 75 | ||
FQ222897.1 | - | 1 - 2 | 247 | 75 | ||
FQ223053.1 | - | 1 - 2 | 245 | 75 | ||
FQ224782.1 | - | 1 - 2 | 246 | 75 | ||
FQ228448.1 | - | 1 - 2 | 245 | 75 | ||
FQ228589.1 | - | 1 - 2 | 246 | 75 | ||
M55534.1 | - | 785 | 75 | |||
S74229.1 | - | 1 - 2 | 209 | 75 | ||
S77138.1 | - | 1 - 2 | 223 | 75 | ||
S77142.1 | - | 1 - 2 | 208 | 75 | ||
X60351.1 | - | 1 - 2 | 215 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |