Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017189.2 | NP_058885.1 | 6 - 7 | 667 | 80 | |
XM_006246676.2 | XP_006246738.1 | 6 - 7 | 745 | 80 | ||
GenBank mRNA | BC088099.1 | - | 6 - 7 | 547 | 7322417 | 80 |
FQ218583.1 | - | 4 - 5 | 373 | 80 | ||
J02762.1 | - | 6 - 7 | 667 | 80 | ||
M16347.1 | - | 6 - 7 | 646 | 80 | ||
X07636.1 | - | 5 - 6 | 570 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |