Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021696.2 | NP_067728.1 | 6 - 7 | 872 | 99 | |
XM_006249639.2 | XP_006249701.1 | 7 - 8 | 1058 | 99 | ||
XM_008769495.1 | XP_008767717.1 | 7 - 8 | 955 | 99 | ||
GenBank mRNA | X64563.1 | - | 6 - 7 | 873 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |