Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021744.1 | NP_068512.1 | 1 - 2 | 67 | 56 | |
GenBank mRNA | AF087943.1 | - | 1 - 2 | 67 | 56 | |
BC061733.1 | - | 1 - 2 | 205 | 5598794 | 56 | |
FQ219480.1 | - | 1 - 2 | 100 | 56 | ||
U51804.1 | - | 1 - 2 | 89 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |