Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021771.2 | NP_068539.2 | 5 - 6 | 1546 | 70 | |
XM_006232282.2 | XP_006232344.2 | 5 - 6 | 1711 | 70 | ||
XM_008760941.1 | XP_008759163.1 | 5 - 6 | 1711 | 70 | ||
GenBank mRNA | JN160741.1 | - | 5 - 6 | 1307 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |