Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022248.1 | NP_071584.1 | 5 - 6 | 875 | 73 | |
GenBank mRNA | AF217233.1 | - | 5 - 6 | 875 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |