Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022284.2 | NP_071620.1 | 2 - 3 | 298 | 70 | |
GenBank mRNA | U41322.1 | - | 2 - 3 | 298 | 70 | |
U73898.1 | - | 2 - 3 | 314 | 70 | ||
U75186.1 | - | 2 - 3 | 425 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |