Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022604.2 | NP_072126.1 | 2 - 3 | 589 | 89 | |
GenBank mRNA | BC070888.1 | - | 2 - 3 | 589 | 7098029 | 89 |
U80818.1 | - | 2 - 3 | 502 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |