Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022703.3 | NP_073194.1 | 6 - 7 | 563 | 65 | |
XM_006240996.1 | XP_006241058.1 | 6 - 7 | 668 | 65 | ||
XM_006240997.1 | XP_006241059.1 | 6 - 7 | 665 | 65 | ||
XM_006240998.2 | XP_006241060.1 | 6 - 7 | 2508 | 65 | ||
GenBank mRNA | AF368278.1 | - | 6 - 7 | 575 | 65 | |
AJ222724.1 | - | 6 - 7 | 551 | 65 | ||
BC087642.1 | - | 6 - 7 | 563 | 7308544 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |