Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022703.3 | NP_073194.1 | 9 - 10 | 799 | 76 | |
XM_006240996.1 | XP_006241058.1 | 9 - 10 | 904 | 76 | ||
XM_006240997.1 | XP_006241059.1 | 9 - 10 | 901 | 76 | ||
XM_006240998.2 | XP_006241060.1 | 9 - 10 | 2744 | 76 | ||
GenBank mRNA | AF368278.1 | - | 9 - 10 | 811 | 76 | |
AJ222724.1 | - | 9 - 10 | 787 | 76 | ||
BC087642.1 | - | 9 - 10 | 799 | 7308544 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |