Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022962.1 | NP_075251.1 | 1 - 2 | 191 | 125 | |
GenBank mRNA | AF081144.1 | - | 1 - 2 | 191 | 125 | |
AF081145.1 | - | 1 - 2 | 191 | 125 | ||
AF081146.1 | - | 1 - 2 | 191 | 125 | ||
AF081147.1 | - | 1 - 2 | 191 | 125 | ||
AF111099.1 | - | 1 - 2 | 367 | 125 | ||
U78105.1 | - | 1 - 2 | 333 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Inventoried Probe spans exons |