Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024386.1 | NP_077362.1 | 1 - 2 | 75 | 94 | |
GenBank mRNA | BC061797.1 | - | 1 - 2 | 77 | 5598033 | 94 |
FQ209392.1 | - | 1 - 2 | 73 | 94 | ||
FQ209616.1 | - | 1 - 2 | 79 | 94 | ||
FQ234382.1 | - | 1 - 2 | 73 | 94 | ||
Y10054.1 | - | 1 - 2 | 75 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |