Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031796.1 | NP_113984.1 | 3 - 4 | 1818 | 87 | |
GenBank mRNA | AF049344.1 | - | 3 - 4 | 1818 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |