Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3ar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032060.1 | NP_114449.1 | 1 - 2 | 123 | 75 | |
XM_006237314.2 | XP_006237376.1 | 1 - 2 | 363 | 75 | ||
GenBank mRNA | U86379.1 | - | 1 - 2 | 123 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus Inventoried Probe spans exons |