Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052801.1 | NP_434688.1 | 1 - 2 | 352 | 79 | |
GenBank mRNA | S80345.1 | - | 1 - 2 | 293 | 79 | |
U14746.1 | - | 1 - 2 | 352 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Rat Chemokine Panel Rat Hypoxia Panel 1700 Inventoried Probe spans exons |