Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053325.1 | NP_445777.1 | 2 - 3 | 286 | 63 | |
GenBank mRNA | AF375462.1 | - | 2 - 3 | 286 | 63 | |
AF375467.1 | - | 2 - 3 | 286 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |