Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053373.1 | NP_445825.1 | 2 - 3 | 392 | 60 | |
GenBank mRNA | AF154114.1 | - | 2 - 3 | 392 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |