Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053634.1 | NP_446086.1 | 7 - 8 | 603 | 131 | |
GenBank mRNA | AB036792.1 | - | 7 - 8 | 603 | 131 | |
BC088123.1 | - | 7 - 8 | 595 | 7376834 | 131 | |
FQ220557.1 | - | 7 - 8 | 606 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |