Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270939.1 | NP_001257868.1 | 2 - 3 | 151 | 62 | |
NM_053969.3 | NP_446421.3 | 2 - 3 | 325 | 62 | ||
XM_006247865.1 | XP_006247927.1 | 2 - 3 | 320 | 62 | ||
XM_006247866.1 | XP_006247928.1 | 2 - 3 | 320 | 62 | ||
XM_006247867.2 | XP_006247929.1 | 2 - 3 | 180 | 62 | ||
XM_008768462.1 | XP_008766684.1 | 2 - 3 | 320 | 62 | ||
XM_008768463.1 | XP_008766685.1 | 2 - 3 | 180 | 62 | ||
GenBank mRNA | BC061746.1 | - | 2 - 3 | 274 | 5599712 | 62 |
CB792560.1 | - | 2 - 3 | 201 | 62 | ||
X87885.1 | - | 1 - 2 | 106 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |