Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057109.1 | NP_476450.1 | 2 - 3 | 1454 | 91 | |
GenBank mRNA | AB043981.1 | - | 2 - 3 | 1454 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |