Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133314.1 | NP_579848.1 | 2 - 3 | 1044 | 82 | |
GenBank mRNA | AF238865.1 | - | 2 - 3 | 1043 | 82 | |
AF335281.1 | - | 2 - 3 | 1044 | 82 | ||
BC085696.1 | - | 1065 | 7190374 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |