Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133416.1 | NP_596907.1 | 1 - 2 | 439 | 76 | |
GenBank mRNA | AF378332.1 | - | 1 - 2 | 439 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |