Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138851.1 | NP_620206.1 | 2 - 3 | 195 | 96 | |
GenBank mRNA | AY089983.1 | - | 2 - 3 | 195 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |