Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153476.1 | NP_703206.1 | 1 - 2 | 629 | 127 | |
GenBank mRNA | AY128946.1 | - | 1 - 2 | 629 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |