Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153626.1 | NP_705890.1 | 1 - 2 | 209 | 109 | |
XM_006230697.2 | XP_006230759.1 | 3 - 4 | 2366 | 109 | ||
GenBank mRNA | AB050103.1 | - | 1 - 2 | 209 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |