Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019358.1 | NP_062231.1 | 2 - 3 | 434 | 83 | |
XM_006239338.2 | XP_006239400.1 | 2 - 3 | 434 | 83 | ||
GenBank mRNA | BC072492.1 | - | 2 - 3 | 390 | 7097186 | 83 |
U07797.1 | - | 2 - 3 | 434 | 83 | ||
U32115.1 | - | 2 - 3 | 362 | 83 | ||
U96449.1 | - | 2 - 3 | 214 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |