Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020656.2 | NP_065707.2 | 2 - 3 | 222 | 72 | |
GenBank mRNA | AF264765.2 | - | 2 - 3 | 222 | 72 | |
FQ228521.1 | - | 282 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |