Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034198.2 | NP_001029370.1 | 3 - 4 | 306 | 63 | |
NM_021671.3 | NP_067703.1 | 3 - 4 | 306 | 63 | ||
GenBank mRNA | AB006135.1 | - | 3 - 4 | 264 | 63 | |
CK359233.1 | - | 3 - 4 | 187 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |