Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022008.1 | NP_071291.1 | 2 - 3 | 154 | 69 | |
XM_006228843.2 | XP_006228905.1 | 2 - 3 | 234 | 69 | ||
GenBank mRNA | AW914994.1 | - | 2 - 3 | 154 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |