Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030585.1 | NP_085074.1 | 8 - 9 | 1263 | 102 | |
GenBank mRNA | BC089795.1 | - | 8 - 9 | 1249 | 7375879 | 102 |
U34932.1 | - | 8 - 9 | 1263 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |