Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031062.2 | NP_112324.1 | 8 - 9 | 1059 | 76 | |
GenBank mRNA | BC081784.1 | - | 8 - 9 | 1044 | 7135885 | 76 |
U53706.1 | - | 8 - 9 | 1060 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |