Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022525.3 | NP_071970.2 | 3 - 4 | 424 | 77 | |
GenBank mRNA | BC062227.1 | - | 3 - 4 | 400 | 6921653 | 77 |
CK480346.1 | - | 3 - 4 | 424 | 77 | ||
CV079715.1 | - | 2 - 3 | 163 | 77 | ||
D00680.1 | - | 3 - 4 | 489 | 77 | ||
FQ221037.1 | - | 3 - 4 | 396 | 77 | ||
FQ221279.1 | - | 3 - 4 | 377 | 77 | ||
FQ221454.1 | - | 3 - 4 | 396 | 77 | ||
FQ223139.1 | - | 2 - 3 | 139 | 77 | ||
FQ228485.1 | - | 3 - 4 | 396 | 77 | ||
FQ228545.1 | - | 3 - 4 | 450 | 77 | ||
FQ228679.1 | - | 3 - 4 | 396 | 77 | ||
FQ228690.1 | - | 3 - 4 | 409 | 77 | ||
FQ228694.1 | - | 3 - 4 | 376 | 77 | ||
FQ228697.1 | - | 3 - 4 | 396 | 77 | ||
FQ228699.1 | - | 3 - 4 | 396 | 77 | ||
FQ228705.1 | - | 3 - 4 | 394 | 77 | ||
FQ228712.1 | - | 3 - 4 | 396 | 77 | ||
FQ228719.1 | - | 3 - 4 | 396 | 77 | ||
FQ228737.1 | - | 3 - 4 | 396 | 77 | ||
FQ228751.1 | - | 3 - 4 | 396 | 77 | ||
FQ228802.1 | - | 3 - 4 | 396 | 77 | ||
FQ228803.1 | - | 3 - 4 | 393 | 77 | ||
FQ228804.1 | - | 3 - 4 | 396 | 77 | ||
FQ228807.1 | - | 3 - 4 | 397 | 77 | ||
FQ228808.1 | - | 3 - 4 | 396 | 77 | ||
FQ228824.1 | - | 3 - 4 | 396 | 77 | ||
FQ228835.1 | - | 3 - 4 | 396 | 77 | ||
FQ228845.1 | - | 3 - 4 | 396 | 77 | ||
FQ228878.1 | - | 3 - 4 | 396 | 77 | ||
FQ228879.1 | - | 3 - 4 | 396 | 77 | ||
FQ228886.1 | - | 3 - 4 | 396 | 77 | ||
FQ228899.1 | - | 3 - 4 | 396 | 77 | ||
FQ228914.1 | - | 3 - 4 | 396 | 77 | ||
FQ228921.1 | - | 3 - 4 | 396 | 77 | ||
FQ228934.1 | - | 3 - 4 | 398 | 77 | ||
FQ228945.1 | - | 3 - 4 | 396 | 77 | ||
FQ228947.1 | - | 3 - 4 | 396 | 77 | ||
FQ228960.1 | - | 3 - 4 | 396 | 77 | ||
FQ228965.1 | - | 3 - 4 | 396 | 77 | ||
FQ228970.1 | - | 3 - 4 | 396 | 77 | ||
FQ228976.1 | - | 3 - 4 | 396 | 77 | ||
FQ229118.1 | - | 3 - 4 | 396 | 77 | ||
FQ229125.1 | - | 3 - 4 | 396 | 77 | ||
FQ230052.1 | - | 2 - 3 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |