Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031977.1 | NP_114183.1 | 6 - 7 | 714 | 56 | |
XM_008762387.1 | XP_008760609.1 | 6 - 7 | 723 | 56 | ||
XM_008762388.1 | XP_008760610.1 | 6 - 7 | 760 | 56 | ||
XM_008762389.1 | XP_008760611.1 | 6 - 7 | 831 | 56 | ||
GenBank mRNA | AF130457.1 | - | 5 - 6 | 696 | 56 | |
AF157016.1 | - | 6 - 7 | 714 | 56 | ||
DQ120509.1 | - | 5 - 6 | 680 | 56 | ||
DQ120510.1 | - | 5 - 6 | 680 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus Probe spans exons |