Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053582.2 | NP_446034.1 | 3 - 4 | 470 | 75 | |
XM_006238977.2 | XP_006239039.1 | 4 - 5 | 562 | 75 | ||
XM_006238978.2 | XP_006239040.1 | 4 - 5 | 559 | 75 | ||
GenBank mRNA | AB050717.1 | - | 3 - 4 | 471 | 75 | |
BC097299.1 | - | 4 - 5 | 529 | 7445067 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |