Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053595.2 | NP_446047.1 | 3 - 4 | 605 | 77 | |
GenBank mRNA | BC087006.1 | - | 3 - 4 | 605 | 7193034 | 77 |
L40030.1 | - | 3 - 4 | 340 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |