Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013197.1 | NP_037329.1 | 1 - 2 | 193 | 85 | |
GenBank mRNA | AF186775.1 | - | 236 | 85 | ||
BC081857.1 | - | 214 | 7133382 | 85 | ||
D86297.1 | - | 1 - 2 | 193 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |