Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133313.2 | NP_579847.1 | 7 - 8 | 988 | 78 | |
GenBank mRNA | AJ301634.1 | - | 7 - 8 | 1097 | 78 | |
BC062087.1 | - | 7 - 8 | 988 | 5621521 | 78 | |
FQ213088.1 | - | 2 - 3 | 220 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |