Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138855.1 | NP_620210.1 | 4 - 5 | 459 | 142 | |
XM_006239766.1 | XP_006239828.1 | 4 - 5 | 693 | 142 | ||
XM_006239767.2 | XP_006239829.1 | 4 - 5 | 657 | 142 | ||
XM_006239768.2 | XP_006239830.1 | 4 - 5 | 524 | 142 | ||
XM_008764502.1 | XP_008762724.1 | 4 - 5 | 1359 | 142 | ||
GenBank mRNA | AF492385.1 | - | 4 - 5 | 459 | 142 | |
BC078729.1 | - | 3 - 4 | 382 | 7134092 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |