Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145773.1 | NP_665716.1 | 1 - 2 | 146 | 65 | |
GenBank mRNA | FQ234290.1 | - | 1 - 2 | 136 | 65 | |
U45986.1 | - | 1 - 2 | 146 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |