Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012728.1 | NP_036860.1 | 1 - 2 | 93 | 88 | |
GenBank mRNA | M97797.1 | - | 1 - 2 | 93 | 88 | |
S75952.1 | - | 1 - 2 | 87 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |