Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012879.2 | NP_037011.2 | 5 - 6 | 636 | 66 | |
XM_006232207.2 | XP_006232269.1 | 6 - 7 | 940 | 66 | ||
GenBank mRNA | BC078875.1 | - | 5 - 6 | 636 | 7127751 | 66 |
J03145.1 | - | 5 - 6 | 711 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |