Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190236.1 | NP_001177165.1 | 12 - 13 | 1530 | 70 | |
NM_001190237.1 | NP_001177166.1 | 12 - 13 | 1527 | 70 | ||
NM_001190238.1 | NP_001177167.1 | 11 - 12 | 1518 | 70 | ||
NM_013013.2 | NP_037145.2 | 11 - 12 | 1521 | 70 | ||
XM_006256403.1 | XP_006256465.1 | 12 - 13 | 1539 | 70 | ||
GenBank mRNA | BC061759.1 | - | 11 - 12 | 1452 | 5600088 | 70 |
DY313840.1 | - | 2 - 3 | 386 | 70 | ||
FQ217536.1 | - | 11 - 12 | 1472 | 70 | ||
M19936.1 | - | 11 - 12 | 1471 | 70 | ||
S81353.1 | - | 11 - 12 | 1471 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |