Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016988.2 | NP_058684.2 | 5 - 6 | 588 | 69 | |
XM_006234500.1 | XP_006234562.1 | 6 - 7 | 1510 | 69 | ||
GenBank mRNA | BC081823.1 | - | 5 - 6 | 559 | 7107570 | 69 |
M27893.1 | - | 5 - 6 | 557 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |