Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270889.1 | NP_001257818.1 | 2 - 3 | 412 | 62 | |
NM_019144.2 | NP_062017.2 | 2 - 3 | 354 | 62 | ||
XM_006242691.2 | XP_006242753.1 | 4 - 5 | 1060 | 62 | ||
XM_006242692.2 | XP_006242754.1 | 3 - 4 | 1005 | 62 | ||
XM_006242693.2 | XP_006242755.1 | 2 - 3 | 871 | 62 | ||
XM_006242694.2 | XP_006242756.1 | 2 - 3 | 308 | 62 | ||
GenBank mRNA | BC078847.1 | - | 2 - 3 | 412 | 7110955 | 62 |
CV074238.1 | - | 2 - 3 | 354 | 62 | ||
M76110.1 | - | 2 - 3 | 352 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |