Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024155.3 | NP_077069.3 | 5 - 6 | 389 | 90 | |
XM_006236841.2 | XP_006236903.1 | 6 - 7 | 476 | 90 | ||
XM_006236842.2 | XP_006236904.1 | 5 - 6 | 1012 | 90 | ||
GenBank mRNA | BC085688.1 | - | 5 - 6 | 389 | 7191458 | 90 |
D38224.2 | - | 3 - 4 | 298 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |