Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082479.1 | NP_001075948.1 | 3 - 4 | 679 | 68 | |
NM_178866.4 | NP_849197.3 | 3 - 4 | 382 | 68 | ||
XM_006241193.2 | XP_006241255.1 | 4 - 5 | 522 | 68 | ||
GenBank mRNA | BC086374.1 | - | 3 - 4 | 382 | 7300903 | 68 |
D00698.1 | - | 3 - 4 | 363 | 68 | ||
FQ209824.1 | - | 4 - 5 | 511 | 68 | ||
FQ210688.1 | - | 3 - 4 | 395 | 68 | ||
FQ225388.1 | - | 3 - 4 | 395 | 68 | ||
FQ225437.1 | - | 3 - 5 | 68 | |||
FQ231951.1 | - | 3 - 4 | 395 | 68 | ||
FQ234943.1 | - | 3 - 4 | 395 | 68 | ||
M15481.1 | - | 3 - 4 | 1114 | 68 | ||
M17335.1 | - | 3 - 4 | 579 | 68 | ||
X06043.1 | - | 2 - 3 | 264 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |