Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp26b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181087.2 | NP_851601.1 | 4 - 5 | 1019 | 66 | |
GenBank mRNA | AY245532.2 | - | 4 - 5 | 1019 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |