Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smagp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182817.1 | NP_877969.1 | 3 - 4 | 173 | 60 | |
XM_006242323.2 | XP_006242385.1 | 3 - 4 | 302 | 60 | ||
GenBank mRNA | AJ577837.1 | - | 3 - 4 | 173 | 60 | |
BC101910.1 | - | 3 - 4 | 488 | 7933976 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |