Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017257.2 | NP_058953.1 | 6 - 7 | 361 | 105 | |
GenBank mRNA | BC058486.1 | - | 6 - 7 | 399 | 6918985 | 105 |
BC097331.1 | - | 6 - 7 | 361 | 7389180 | 105 | |
D45250.1 | - | 6 - 7 | 411 | 105 | ||
FQ226805.1 | - | 6 - 7 | 392 | 105 | ||
FQ227049.1 | - | 6 - 7 | 405 | 105 | ||
FQ232840.1 | - | 6 - 7 | 391 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |