Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021846.2 | NP_068618.2 | 947 | 64 | ||
GenBank mRNA | AF115380.1 | - | 879 | 64 | ||
AF144096.1 | - | 203 | 64 | |||
BC078835.1 | - | 947 | 7121699 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |